Skip to Content
Merck
All Photos(1)

Key Documents

275573

Sigma-Aldrich

4-Hydroxybutyl acrylate

90%, contains 50 ppm monomethyl ether hydroquinone as inhibitor, 300 ppm hydroquinone as inhibitor

Synonym(s):

1,4-Butanediol acrylate, Tetramethylene glycol monoacrylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CHCO2(CH2)4OH
CAS Number:
Molecular Weight:
144.17
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

90%

contains

300 ppm hydroquinone as inhibitor
50 ppm monomethyl ether hydroquinone as inhibitor

refractive index

n20/D 1.452 (lit.)

bp

95 °C/0.1 mmHg (lit.)

density

1.041 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

OCCCCOC(=O)C=C

InChI

1S/C7H12O3/c1-2-7(9)10-6-4-3-5-8/h2,8H,1,3-6H2

InChI key

NDWUBGAGUCISDV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Hydroxybutyl acrylate (HBA) can be synthesized by the esterification of acrylic acid with 1, 4 butanediol over a catalyst such as Amberlyst 15. 4-Hydroxybutyl acrylate (HBA) exhibits properties such as luster, chemical and scratch resistance. HBA monomers may be used as one of the constituents for the fabrication of a membrane system for controlled release of transdermal drug delivery system.

Application

Hydroxybutyl acrylate was one of the monomers utilized in the preparation of poly(2-hydroxy-3-phenoxypropylacrylate, 4-hydroxybutyl acrylate, dibutyl maleate) membranes.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Agnieszka Kowalczyk et al.
Polymers, 11(12) (2019-12-15)
New organic-inorganic hybrid copolymers (EA-POSSs) based on butyl acrylate, glycidyl methacylate, hydroxybutyl acrylate, acryloiloxybenzophenone and acryloxypropyl-heptaisobutyl-POSS (A-POSS) were prepared via free-radical solution polymerization (FRP) and applied as a component of thermally curable structural self-adhesive tapes (SATs). The EA-POSS with 0.25
A new poly(2-hydroxy-3-phenoxypropylacrylate, 4-hydroxybutyl acrylate, diethyl maleate) membrane controlled clonidine linear release in the transdermal drug delivery system.
Zhan X, et al.
European Polymer Journal, 43(4), 1588-1594 (2007)
Qian Zhao et al.
Journal of materials chemistry. B, 8(40), 9362-9373 (2020-10-01)
A series of novel nanofibrillated cellulose (NFC) reinforced gradient intelligent hydrogels with high response rate, multiple response patterns and diversified self-driven functions were successfully prepared. Based on the effect of the hydroxide radical of NFC on the addition reaction, and
A new poly(2-hydroxy-3-phenoxypropylacrylate, 4-hydroxybutyl acrylate, diethyl maleate) membrane controlled clonidine linear release in the transdermal drug delivery system.
Zhan X< et al.
European Polymer Journal, 43(4), 83-91 (2007)
Xiaoping Zhan et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 66(3), 429-434 (2007-01-02)
Poly(2-hydroxy-3-phenoxypropylacrylate, 4-hydroxybutyl acrylate, dibutyl maleate) membrane was synthesized by UV curing method in our laboratory for the first time. When above-mentioned monomers were in the weight ratio of 4:4:2, the membrane not only had perfect permeation property but also had

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service