Skip to Content
Merck
All Photos(2)

Key Documents

230669

Sigma-Aldrich

4-Phenoxyphenol

99%

Synonym(s):

Hydroquinone monophenyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5OC6H4OH
CAS Number:
Molecular Weight:
186.21
Beilstein:
2047182
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

99%

form

solid

mp

80-84 °C (lit.)

functional group

phenoxy

SMILES string

Oc1ccc(Oc2ccccc2)cc1

InChI

1S/C12H10O2/c13-10-6-8-12(9-7-10)14-11-4-2-1-3-5-11/h1-9,13H

InChI key

ZSBDGXGICLIJGD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

338.0 °F - closed cup

Flash Point(C)

170 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Victoria B F Custodis et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 23(36), 8658-8668 (2017-04-08)
One of the key challenges in renewable chemical production is the conversion of lignin, especially by fast pyrolysis. The complexity of the lignin pyrolysis process has hindered the elucidation of the mechanism, inhibiting further industrial implementation. By combining pyrolysis of
Kyoungseon Min et al.
Biotechnology for biofuels, 10, 212-212 (2017-09-16)
In the biorefinery utilizing lignocellulosic biomasses, lignin decomposition to value-added phenolic derivatives is a key issue, and recently biocatalytic delignification is emerging owing to its superior selectivity, low energy consumption, and unparalleled sustainability. However, besides heme-containing peroxidases and laccases, information
Xiaolu Jiang et al.
Bioorganic & medicinal chemistry letters, 18(24), 6549-6552 (2008-10-28)
The synthesis and biological evaluation of a series of diphenyl ether derivatives were described. The compounds can either activate or inhibit the aminopeptidase activity of leukotriene A(4) hydrolase, while at the same time do not influence the hydrolase activity. Further
Cynthia D Selassie et al.
Journal of medicinal chemistry, 48(23), 7234-7242 (2005-11-11)
In this comprehensive study on the caspase-mediated apoptosis-inducing effect of 51 substituted phenols in a murine leukemia cell line (L1210), we determined the concentrations needed to induce caspase activity by 50% (I50) and utilized these data to develop the following

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service