Skip to Content
Merck
All Photos(4)

Key Documents

228982

Sigma-Aldrich

Benzyltrimethylammonium chloride

97%

Synonym(s):

Amberlite IRC 178(Cl), Benzyltrimethylammonium chloride (6CI), Trimethylbenzylammonium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2N(Cl)(CH3)3
CAS Number:
Molecular Weight:
185.69
Beilstein:
3917255
EC Number:
MDL number:
UNSPSC Code:
12352107
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

97%

mp

239 °C (dec.) (lit.)

SMILES string

[Cl-].C[N+](C)(C)Cc1ccccc1

InChI

1S/C10H16N.ClH/c1-11(2,3)9-10-7-5-4-6-8-10;/h4-8H,9H2,1-3H3;1H/q+1;/p-1

InChI key

KXHPPCXNWTUNSB-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Benzyltrimethylammonium chloride can be used as a phase catalyst in:
  • The synthesis of alkyl and aryl sulfonamides from thiols and disulfides in the presence of 1,3-dichloro-5,5-dimethylhydantoin (DCH).[1]
  • The selective oxidation of benzyl alcohol to benzaldehyde using hydrogen peroxide as oxidant.[2]
  • The esterification reaction of ethanol and lauric acid.[3]

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Muta. 2

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kinetics of the reaction of ethanol and lauric acid catalyzed by deep eutectic solvent based on benzyltrimethylammonium chloride
Li Kun, et al.
The Canadian Journal of Chemical Engineering, 97(5), 1144-1151 (2019)
Selective oxidation of benzyl alcohol with hydrogen peroxide over reaction-controlled phase-transfer catalyst
Weng Z, et al.
Catalysis Communications, 8(10), 1493-1496 (2007)
Convenient one-pot Synthesis of sulfonamides from thiols and disulfides using 1, 3-dichloro-5, 5-dimethylhydantoin (DCH)
Veisi Hojat
Bull. Korean Chem. Soc., 33(2), 383-386 (2012)
Kathrin Müller et al.
Analytical and bioanalytical chemistry, 412(20), 4867-4879 (2020-03-05)
Matrix effects have been shown to be very pronounced and highly variable in the analysis of mobile chemicals, which may severely exacerbate accurate quantification. These matrix effects, however, are still scarcely studied in combination with hydrophilic interaction liquid chromatography (HILIC)
L Sangeetha Vedula et al.
Archives of biochemistry and biophysics, 466(2), 260-266 (2007-08-07)
Trichodiene synthase is a terpenoid cyclase that catalyzes the cyclization of farnesyl diphosphate (FPP) to form the bicyclic sesquiterpene hydrocarbon trichodiene (89%), at least five sesquiterpene side products (11%), and inorganic pyrophosphate (PP(i)). Incubation of trichodiene synthase with 2-fluorofarnesyl diphosphate

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service