Skip to Content
Merck
All Photos(1)

Key Documents

216704

Sigma-Aldrich

cis-5-Norbornene-endo-2,3-dicarboxylic acid

98%

Synonym(s):

NDC

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H10O4
CAS Number:
Molecular Weight:
182.17
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Quality Level

Assay

98%

mp

175 °C (dec.) (lit.)

SMILES string

OC(=O)[C@@H]1[C@H]2C[C@H](C=C2)[C@@H]1C(O)=O

InChI

1S/C9H10O4/c10-8(11)6-4-1-2-5(3-4)7(6)9(12)13/h1-2,4-7H,3H2,(H,10,11)(H,12,13)/t4-,5+,6-,7+

InChI key

NIDNOXCRFUCAKQ-UMRXKNAASA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Malcolm Driffield et al.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 35(6), 1200-1213 (2018-02-24)
The polymeric coating used in metal packaging such as cans for foods and beverages may contain residual amounts of monomers used in the production of the coating, as well as unreacted linear and cyclic oligomers. Traditionally, although designed for use

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service