Skip to Content
Merck
All Photos(1)

Key Documents

189731

Sigma-Aldrich

5-Methyl-2-hexanol

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2CH2CHOHCH3
CAS Number:
Molecular Weight:
116.20
Beilstein:
1731619
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.417 (lit.)

bp

148-150 °C (lit.)

density

0.819 g/mL at 25 °C (lit.)

functional group

hydroxyl

SMILES string

CC(C)CCC(C)O

InChI

1S/C7H16O/c1-6(2)4-5-7(3)8/h6-8H,4-5H2,1-3H3

InChI key

ZDVJGWXFXGJSIU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

5-Methyl-2-hexanol is a non-crystallizable secondary alcohol and its complex relative permittivity has been measured over wide range of temperatures and pressures.

Application

5-Methyl-2-hexanol has been used as internal standard for rapid extraction of aroma compounds from grape brandies and aqueous-alcoholic wood extracts by ultrasound.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

86.0 °F - closed cup

Flash Point(C)

30 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lee Wei Lim et al.
Scientific reports, 6, 37568-37568 (2016-11-22)
Tetratricopeptide repeat domain 9A (TTC9A) expression is abundantly expressed in the brain. Previous studies in TTC9A knockout (TTC9A
Improved method for extraction of aroma compounds in aged brandies and aqueous alcoholic wood extracts using ultrasound.
Caldeira I, et al.
Analytica Chimica Acta, 513(1), 125-134 (2004)
S Pawlus et al.
The Journal of chemical physics, 135(8), 084507-084507 (2011-09-08)
The complex relative permittivity of a non-crystallizable secondary alcohol, 5-methyl-2-hexanol, is measured over a wide range of temperatures and pressures up to 1750 MPa (17.5 kbar). The data at atmospheric pressure (P = 0.101 MPa) are analyzed in terms of
Li Guan et al.
Brain research bulletin, 157, 162-168 (2020-02-15)
The involvement of tetratricopeptide repeat domain 9A (TTC9A) deficiency in anxiety-like responses and behavioral despair through estradiol action on the serotonergic system has been reported. Emerging evidence suggests that estradiol is a potent modulator of neuroplasticity. As estradiol and neuroplasticity
Sijie Tan et al.
Experimental cell research, 382(1), 111433-111433 (2019-05-18)
Autophagy is an evolutionary conserved, self-eating process that targets cellular constituents for lysosomal degradation. Transcription factor EB (TFEB) is a master regulator of autophagy by inducing the expression of genes involved in autophagic and lysosomal degradation. In breast cancer, ligand-activated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service