Skip to Content
Merck
All Photos(2)

Key Documents

182389

Sigma-Aldrich

Polypropylene, isotactic

average Mw ~250,000 by GPC

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH2CH(CH3)]n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

solid

autoignition temp.

>674 °F

mol wt

average Mw ~250,000 by GPC

refractive index

n20/D 1.49 (lit.)

mp

189 °C (lit.)

transition temp

Tg −26 °C

density

0.9 g/mL at 25 °C (lit.)

SMILES string

CC=C

InChI

1S/C3H6/c1-3-2/h3H,1H2,2H3

InChI key

QQONPFPTGQHPMA-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Eric S Kim et al.
Polymers, 11(2) (2019-04-10)
Strain hardening has important roles in understanding material structures and polymer processing methods, such as foaming, film forming, and fiber extruding. A common method to improve strain hardening behavior is to chemically branch polymer structures, which is costly, thus preventing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service