Skip to Content
Merck
All Photos(3)

Key Documents

151467

Sigma-Aldrich

5-Ethylidene-2-norbornene

contains 100-700 ppm BHT as inhibitor, mixture of endo and exo, 99%

Synonym(s):

2-Ethylidene-5-norbornene, 2-Ethylidenebicyclo[2.2.1]hept-5-ene, 5-Ethylidenebicyclo[2.2.1]hept-2-ene, 5-Ethylidenenorbornene, Ethylidenenorbornene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H12
CAS Number:
Molecular Weight:
120.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

4.1 (vs air)

Quality Level

vapor pressure

4.6 mmHg ( 20 °C)

Assay

99%

form

liquid

contains

100-700 ppm BHT as inhibitor

expl. lim.

~6.4 %

refractive index

n20/D 1.49 (lit.)

bp

146 °C (lit.)

density

0.893 g/mL at 25 °C (lit.)

SMILES string

C\C=C1/C[C@H]2C[C@@H]1C=C2

InChI

1S/C9H12/c1-2-8-5-7-3-4-9(8)6-7/h2-4,7,9H,5-6H2,1H3/b8-2+/t7-,9-/m0/s1

InChI key

OJOWICOBYCXEKR-WHZSQGQPSA-N

Looking for similar products? Visit Product Comparison Guide

General description

5-Ethylidene-2-norbornene, also known as ethylidenenorbornene, is a bicyclic diene and a commercial petrochemical. It is widely used in the chemical industry as a copolymer for the synthesis of various polymer materials.

Application

5-Ethylidene-2-norbornene was used as self-healing agent for polymeric composites.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT RE 2 Inhalation

Target Organs

Liver,Testes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

83.3 °F - closed cup

Flash Point(C)

28.5 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lee JK, et al.
Macromolecular Research, 12(5), 2004-2004 (2004)
B Ballantyne et al.
Journal of applied toxicology : JAT, 17(4), 211-221 (1997-07-01)
The acute toxicity and primary irritancy of the industrial chemicals 5-ethylidene-2-norbornene (ENB) and 5-vinyl-2-norbornene (VNB) were studied. They are of moderate acute peroral toxicity in the rat, with LD50 values for ENB of 2.54 (male) and 5.66 (female) ml kg(-1)
B Ballantyne et al.
Journal of applied toxicology : JAT, 17(4), 197-210 (1997-07-01)
5-Ethylidene-2-norbornene (ENB) is an industrial chemical whose physical properties indicate a likelihood for vapor exposure to humans. The potential for target organ or cumulative toxicity was investigated in rats exposed for 6 h per day for 9 days over an
Carolyn Lee et al.
Chemosphere, 56(3), 285-297 (2004-06-03)
Several predictive models were used to assess aquatic exposure, persistence (P) and potential for long-range transport (LRT) of 5-ethylidene-2-norbornene (ENB). Such estimations are components of the assessment process for persistent, bioaccumulative and toxic (PBT) substances, which are also referred to
5-Ethylidene-2-norbornene.
B Ballantyne et al.
Journal of applied toxicology : JAT, 19(4), 291-294 (1999-08-10)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service