Skip to Content
Merck
All Photos(1)

Key Documents

144711

Sigma-Aldrich

2-(Bromomethyl)benzonitrile

≥97%

Synonym(s):

α-Bromo-o-tolunitrile, 2-Cyanobenzyl bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrCH2C6H4CN
CAS Number:
Molecular Weight:
196.04
Beilstein:
742605
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

≥97%

form

solid

mp

72-74 °C (lit.)

functional group

bromo
nitrile

SMILES string

BrCc1ccccc1C#N

InChI

1S/C8H6BrN/c9-5-7-3-1-2-4-8(7)6-10/h1-4H,5H2

InChI key

QGXNHCXKWFNKCG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-(Bromomethyl)benzonitrile reacts with 2H-tetrazole in the presence of KOH to yield 4-[(2H-tetra-zol-2-yl)meth-yl]benzonitrile[1]. It undergoes base-promoted condensation reaction with homophthalic anhydride to yield 6,11-dihydro-5H-indeno[1,2-c]isoquinolin-5-one[2].

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Prakash G Jagtap et al.
Organic letters, 7(9), 1753-1756 (2005-04-23)
[reaction: see text] The synthesis of 6,11-dihydro-5H-indeno[1,2-c]isoquinolin-5-ones from the base-promoted condensation reaction of homophthalic anhydride and 2-(bromomethyl)-benzonitrile and a convenient method for the synthesis of indolo[3,2-c]isoquinolinones are described.
Zheng Xing et al.
Acta crystallographica. Section E, Structure reports online, 64(Pt 2), o445-o445 (2008-01-01)
The title compound, C(9)H(7)N(5), was synthesized by reaction of 4-(bromomethyl)benzonitrile and 2H-tetrazole in the presence of KOH. The relative orientation of the planar tetra-zole ring and the methyl-benzonitrile moiety is (-)-anti-clinal. The crystal packing is dominated by van der Waals
William L Scott et al.
Journal of combinatorial chemistry, 11(1), 14-33 (2008-12-25)
Distributed Drug Discovery (D(3)) proposes solving large drug discovery problems by breaking them into smaller units for processing at multiple sites. A key component of the synthetic and computational stages of D(3) is the global rehearsal of prospective reagents and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service