Skip to Content
Merck
All Photos(1)

Documents

133361

Sigma-Aldrich

2-Naphthalenesulfonyl chloride

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H7ClO2S
CAS Number:
Molecular Weight:
226.68
Beilstein:
641898
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

bp

147.7 °C/0.6 mmHg (lit.)

mp

74-76 °C (lit.)

SMILES string

ClS(=O)(=O)c1ccc2ccccc2c1

InChI

1S/C10H7ClO2S/c11-14(12,13)10-6-5-8-3-1-2-4-9(8)7-10/h1-7H

InChI key

OPECTNGATDYLSS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Naphthalenesulfonyl chloride was employed as capping reagent for primary amine to convert histamine to a selective histamine H3-receptor antagonist. It was used for pre-column derivatization during the high-performance liquid chromatographic assay of neomycin sulfate.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M J Tozer et al.
Bioorganic & medicinal chemistry letters, 9(13), 1825-1830 (1999-07-16)
Histamine was converted to a selective histamine H3-receptor antagonist by capping the primary amine with 2-naphthalenesulfonyl chloride. Higher receptor affinity and lower variability in the data from the various bioassays were achieved with the 2-naphthalensulfonamides of histamine homologues.
K Tsuji et al.
Journal of chromatography, 333(2), 365-380 (1985-10-04)
A normal-phase high-performance liquid chromatographic (HPLC) method has been developed for the assay of spectinomycin hydrochloride and spectinomycin sulfate for detection at 254 nm. The method involves pre-column derivatization of secondary amines of spectinomycin with 2-naphthalenesulfonyl chloride (NSCl) using a
K Tsuji et al.
Journal of chromatography, 369(1), 105-115 (1986-11-07)
A normal phase high-performance liquid chromatographic (HPLC) method has been developed for the assay of neomycin sulfate. The method involves pre-column derivatization with 2-naphthalenesulfonyl chloride (NSCl) followed by extraction in chloroform and chromatography using a normal phase silica column with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service