Skip to Content
Merck
All Photos(1)

Key Documents

124311

Sigma-Aldrich

sec-Butyl sulfide

98%

Synonym(s):

Di-sec-butyl sulfide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[C2H5CH(CH3)]2S
CAS Number:
Molecular Weight:
146.29
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.45 (lit.)

bp

165 °C (lit.)

density

0.839 g/mL at 25 °C (lit.)

SMILES string

CCC(C)SC(C)CC

InChI

1S/C8H18S/c1-5-7(3)9-8(4)6-2/h7-8H,5-6H2,1-4H3

InChI key

IEBAOPMEYUWQPD-UHFFFAOYSA-N

Application

sec-Butyl sulfide was used in the determination of dialkyl sulfides using novel micropacked column prepared from the slurry of bis[1,2-bis(4-n-undecyloxyphenyl)ethane-1,2-dithiolene]nickel(II) coated on chromosorb W by micropacked gas chromatography.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

102.2 °F - closed cup

Flash Point(C)

39 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chiann-Tyng Chou et al.
Journal of chromatography. A, 1043(2), 255-263 (2004-08-28)
The preparation and mesomorphic properties of a substituted bis(dithiolene)nickel complex derived from 4, 4'-dimethoxybenzil are reported. The phase transition temperatures were based on data obtained by polarized light microscopy and differential scanning calorimetry. The mesogenic phase existed over the temperature

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service