Skip to Content
Merck
All Photos(1)

Key Documents

11240

Sigma-Aldrich

DL-Aspartic acid potassium salt hemihydrate

≥98.0% (NT)

Synonym(s):

Potassium DL-aspartate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOOCCH(NH2)CH2COOK · 0.5H2O
CAS Number:
Molecular Weight:
180.20
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26
Pricing and availability is not currently available.

Quality Level

Assay

≥98.0% (NT)

form

powder or crystals

reaction suitability

reaction type: solution phase peptide synthesis

color

white

application(s)

peptide synthesis

SMILES string

O.[K+].[K+].NC(CC(O)=O)C([O-])=O.NC(CC(O)=O)C([O-])=O

InChI

1S/2C4H7NO4.2K.H2O/c2*5-2(4(8)9)1-3(6)7;;;/h2*2H,1,5H2,(H,6,7)(H,8,9);;;1H2/q;;2*+1;/p-2

InChI key

YKZPPPNXRZHVGX-UHFFFAOYSA-L

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service