Skip to Content
Merck
All Photos(4)

Documents

112208

Sigma-Aldrich

4-Nitrobenzoyl chloride

98%

Synonym(s):

p-Nitrobenzoyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4COCl
CAS Number:
Molecular Weight:
185.56
Beilstein:
473192
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

solid

bp

202-205 °C/105 mmHg (lit.)

mp

71-74 °C (lit.)

SMILES string

[O-][N+](=O)c1ccc(cc1)C(Cl)=O

InChI

1S/C7H4ClNO3/c8-7(10)5-1-3-6(4-2-5)9(11)12/h1-4H

InChI key

SKDHHIUENRGTHK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

4-Nitrobenzoyl chloride reacts with triphenylphosphonium salt to synthesize benzofurans (potential positron emission tomography (PET) tracers) by the intramolecular Wittig reaction.It has been used for the derivatisation of Adiol (Androstenediol, an endogenous proliferation agent of prostate cancer).

Application

<ul>
<li><strong>Automated Synthesis of [11C]PiB:</strong> 4-Nitrobenzoyl chloride is utilized in the automated synthesis of [11C]PiB, a radiopharmaceutical for PET imaging of amyloid, highlighting its role in enhancing neuroimaging techniques (Singh et al., 2024).</li>
</ul>

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

215.6 °F - closed cup

Flash Point(C)

102 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Masahiro Ono et al.
Journal of medicinal chemistry, 49(9), 2725-2730 (2006-04-28)
A novel series of benzofuran derivatives as potential positron emission tomography (PET) tracers targeting amyloid plaques in Alzheimer's disease (AD) were synthesized and evaluated. The syntheses of benzofurans were successfully achieved by an intramolecular Wittig reaction between triphenylphosphonium salt and
Tatsuya Higashi et al.
Steroids, 71(11-12), 1007-1013 (2006-09-19)
Androstenediol (Adiol, androst-5-ene-3beta,17beta-diol) is suspected of being an endogenous proliferation agent of prostate cancer (PCa) even after androgen deprivation therapy (ADT). A liquid chromatography-electron capture atmospheric pressure chemical ionization-mass spectrometric (LC-ECAPCI-MS) method for the determination of Adiol in prostatic tissue
Meng-Ge Huangfu et al.
Nanomaterials (Basel, Switzerland), 11(2) (2021-03-07)
The rapid development of advanced high-frequency mobile communication techniques has advanced urgent requirements for polymer materials with high-temperature resistance and good dielectric properties, including low dielectric constants (low-Dk) and low dielectric dissipation factors (low-Df). The relatively poor dielectric properties of
[Experimental establishment of the maximum permissible concentration of para-nitrobenzoylchloride in the air of a work area].
M Ia Kudria et al.
Gigiena truda i professional'nye zabolevaniia, (9)(9), 47-48 (1984-09-01)
Tatsuya Higashi et al.
Steroids, 71(7), 609-617 (2006-04-20)
The liquid chromatography-mass spectrometry (LC-MS) methods were developed and validated for the determination of testosterone (T) in the brain and serum of rats and of 5alpha-androstane-3alpha,17beta-diol (ADIOL), a metabolite of T, in the brain of rats. After derivatization of T

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service