GC Analysis of 4-Methyloctanoic Acid Enantiomers on γ-DEX™ 120
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
γ-DEX 120, 30 m × 0.25 mm I.D., 0.25 μm (24307)
oven
115 °C
inj. temp.
220 °C
detector
FID, 300 °C
carrier gas
helium, 20 cm/sec
injection
1 μL, 100:1 split
sample
~1 mg/mL each enantiomer in methylene chloride
Description
Legal Information
DEX is a trademark of Sigma-Aldrich Co. LLC