Merck
  • STING Pathway Activation Stimulates Potent Immunity against Acute Myeloid Leukemia.
STING Pathway Activation Stimulates Potent Immunity against Acute Myeloid Leukemia.
Cell reports (2016-06-07)
Emily Curran, Xiufen Chen, Leticia Corrales, Douglas E Kline, Thomas W Dubensky, Priyanka Duttagupta, Marcin Kortylewski, Justin Kline
ABSTRACT
Type I interferon (IFN), essential for spontaneous T cell priming against solid tumors, is generated through recognition of tumor DNA by STING. Interestingly, we observe that type I IFN is not elicited in animals with disseminated acute myeloid leukemia (AML). Further, survival of leukemia-bearing animals is not diminished in the absence of type I IFN signaling, suggesting that STING may not be triggered by AML. However, the STING agonist, DMXAA, induces expression of IFN-β and other inflammatory cytokines, promotes dendritic cell (DC) maturation, and results in the striking expansion of leukemia-specific T cells. Systemic DMXAA administration significantly extends survival in two AML models. The therapeutic effect of DMXAA is only partially dependent on host type I IFN signaling, suggesting that other cytokines are important. A synthetic cyclic dinucleotide that also activates human STING provided a similar anti-leukemic effect. These data demonstrate that STING is a promising immunotherapeutic target in AML.
Sigma-Aldrich
ML RR-S2 CDA sodium salt, ≥95% (HPLC)