Merck
  • Synthesis and LC-MS/MS analysis of desmosine-CH2, a potential internal standard for the degraded elastin biomarker desmosine.
Synthesis and LC-MS/MS analysis of desmosine-CH2, a potential internal standard for the degraded elastin biomarker desmosine.
Organic & biomolecular chemistry (2014-10-31)
Yuko Murakami, Rina Suzuki, Hiroto Yanuma, Jiangtao He, Shuren Ma, Gerard M Turino, Yong Y Lin, Toyonobu Usuki
ABSTRACT
Desmosine-CH2, an analog of the elastic tissue degradation biomarker desmosine, can be regarded as a potential internal standard for precise quantification of desmosines by LC-MS/MS. In this study, the chemical synthesis of desmosine-CH2 was completed in 22% overall yield in five steps. The LC-MS/MS analysis of desmosine-CH2 was also achieved.
Sigma-Aldrich
Heptafluorobutyric acid, 98%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Heptafluorobutyric acid, ≥99.0% (GC)
SKU
Pack Size
Availability
Price
Quantity
Supelco
Heptafluorobutyric acid, suitable for ion chromatography, ≥99.5% (GC)
SKU
Pack Size
Availability
Price
Quantity
Supelco
Heptafluorobutyric acid solution, 0.5 M in H2O, LiChropur, suitable for ion pair chromatography