Merck
  • Optimizations of packed sorbent and inlet temperature for large volume-direct aqueous injection-gas chromatography to determine high boiling volatile organic compounds in water.
Optimizations of packed sorbent and inlet temperature for large volume-direct aqueous injection-gas chromatography to determine high boiling volatile organic compounds in water.
Journal of chromatography. A (2014-07-07)
Bofan Yu, Yonghui Song, Lu Han, Huibin Yu, Yang Liu, Hongliang Liu
ABSTRACT
For the expanded application area, fast trace analysis of certain high boiling point (i.e., 150-250 °C) volatile organic compounds (HVOCs) in water, a large volume-direct aqueous injection-gas chromatography (LV-DAI-GC) method was optimized for the following parameters: packed sorbent for sample on-line pretreatment, inlet temperature and detectors configuration. Using the composite packed sorbent self-prepared with lithium chloride and a type of diatomite, the method enabled safe injection of an approximately 50-100 μL sample at an inlet temperature of 150 °C in the splitless mode and separated HVOCs from water matrix in 2 min. Coupled with a flame ionization detector (FID), an electron capture detector (ECD) and a flame photometric detector (FPD), the method could simultaneously quantify 27 HVOCs that belong to seven subclasses (i.e., halogenated aliphatic hydrocarbons, chlorobenzenes, nitrobenzenes, anilines, phenols, polycyclic aromatic hydrocarbons and organic sulfides) in 26 min. Injecting a 50 μL sample without any enrichment step, such as cryotrap focusing, the limits of quantification (LOQs) for the 27 HVOCs was 0.01-3 μg/L. Replicate analyses of the 27 HVOCs spiked source and river water samples exhibited good precision (relative standard deviations ≤ 11.3%) and accuracy (relative errors ≤ 17.6%). The optimized LV-DAI-GC was robust and applicable for fast determination and automated continuous monitoring of HVOCs in surface water.
Sigma-Aldrich
m-Cresol, SAJ first grade, ≥97.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Phenol, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Phenol, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
2-Chlorophenol, SAJ first grade, ≥99.0%
Pricing and availability is not currently available.
Supelco
1,2,3-Trichloropropane, analytical standard
Pricing and availability is not currently available.
Supelco
Phenol solution, 5000 μg/mL in methanol, certified reference material
Pricing and availability is not currently available.
Supelco
Dimethyl trisulfide, analytical standard
Pricing and availability is not currently available.
Supelco
Phenol solution, certified reference material, 500 μg/mL in methanol
Pricing and availability is not currently available.
Sigma-Aldrich
1,2-Dichlorobenzene solution, NMR reference standard, 5% in acetone-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
Pricing and availability is not currently available.
Supelco
Electrolyte solution, nonaqueous, 2 M LiCl in ethanol
Pricing and availability is not currently available.
Sigma-Aldrich
2-Chloroaniline, technical, ≥98.0% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Phenol, JIS special grade, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Phenol, SAJ first grade, ≥98.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Lithium standard solution, suitable for atomic absorption spectrometry, 1000 ppm Li, 1 mg/mL Li
Pricing and availability is not currently available.
Sigma-Aldrich
Nitrobenzene, JIS special grade, ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Phenol, for molecular biology
Pricing and availability is not currently available.
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Phenol solution, BioReagent, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2, for molecular biology
Pricing and availability is not currently available.
Sigma-Aldrich
Phenol solution, BioReagent, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, for molecular biology
Pricing and availability is not currently available.
Supelco
m-Cresol, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
o-Toluidine, purum p.a., ≥99.5% (GC)
Pricing and availability is not currently available.
Supelco
2-Chlorophenol, PESTANAL®, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Phenol, BioUltra, for molecular biology, TE-saturated, ~73% (T)
Pricing and availability is not currently available.
Sigma-Aldrich
Phenol, BioUltra, for molecular biology, ≥99.5% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Pricing and availability is not currently available.
Supelco
Phenol solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
Pricing and availability is not currently available.
Supelco
Phenol, PESTANAL®, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Pricing and availability is not currently available.
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Pricing and availability is not currently available.