Merck
  • Effect of ricinoleic acid esters from castor oil (Ricinus communis) on the oocyte yolk components of the tick Rhipicephalus sanguineus (Latreille, 1806) (Acari: Ixodidae).
Effect of ricinoleic acid esters from castor oil (Ricinus communis) on the oocyte yolk components of the tick Rhipicephalus sanguineus (Latreille, 1806) (Acari: Ixodidae).
Veterinary parasitology (2012-10-09)
Bruno Rodrigues Sampieri, André Arnosti, Karim Christina Scopinho Furquim, Gilberto Orivaldo Chierice, Gervásio Henrique Bechara, Pedro Luiz Pucci Figueiredo de Carvalho, Pablo Henrique Nunes, Maria Izabel Camargo-Mathias
ABSTRACT
Rhipicephalus sanguineus are bloodsucking ectoparasites, whose main host is the domestic dog, thus being present in urban areas and closely located to people. Eventually, this tick species parasitize humans and can become a potential vector of infectious diseases. Methods to control this type of pest have been the focus of many research groups worldwide. The use of natural products is increasingly considered nowadays, due to the low toxicity levels to the host and low waste generation to the environment. This study tested the effect of ricinoleic acid esters from castor oil (as an potential acaricide) on the reproductive system of R. sanguineus females, more specifically on the vitellogenesis process. For this, two groups were established: the control group (CG) and the treatment group (TG) with five rabbits in each (New Zealand White), used as hosts. NaCl and ester were added to rabbits' food and offered to the hosts. After full engorgement, the females were collected and had their ovaries extracted. The ticks ovaries were submitted to histochemical techniques so the effects of esters could be observed over polysaccharides, proteins and lipids yolk. Changes in the deposition of yolk components were observed. This caused modifications on elements of polysaccharide origin and on glycoprotein compounds, interfering in the final yolk synthesis and compromising the development of the future embryo.
Sigma-Aldrich
Castor oil, CP
Sigma-Aldrich
Castor oil
Sigma-Aldrich
Castor oil, meets testing specifications per USP
Sigma-Aldrich
Castor oil, tested according to Ph. Eur.