Merck
  • Pd-catalyzed C3-selective allylation of indoles with allyl alcohols promoted by triethylborane.
Pd-catalyzed C3-selective allylation of indoles with allyl alcohols promoted by triethylborane.
Journal of the American Chemical Society (2005-03-31)
Masanari Kimura, Makoto Futamata, Ryutaro Mukai, Yoshinao Tamaru
ABSTRACT
Under palladium catalysis, Et3B nicely promotes allyl alcohols to undergo C3-selective allylation of indoles and tryptophan; the yields range 75-95%.
Sigma-Aldrich
Triethylborane solution, 1.0 M in THF
Sigma-Aldrich
Triethylborane solution, 1.0 M in hexanes
Sigma-Aldrich
Triethylborane, ≥95%