Skip to Content
Merck
All Photos(1)

Documents

257192

Sigma-Aldrich

Triethylborane

≥95%

Synonym(s):

Triethylboron

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)3B
CAS Number:
Molecular Weight:
97.99
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95%

reaction suitability

reagent type: reductant

refractive index

n20/D 1.397 (lit.)

bp

95 °C (lit.)

mp

−93 °C (lit.)

density

0.677 g/mL at 25 °C (lit.)

SMILES string

CCB(CC)CC

InChI

1S/C6H15B/c1-4-7(5-2)6-3/h4-6H2,1-3H3

InChI key

LALRXNPLTWZJIJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Triethylborane (Et3B) is an organoborane pyrophoric liquid. It is prepared on the plant scale by the reaction of AlEt3 and KBF4. Et3B is widely used as a precursor for the preparation of reducing agents such as lithium triethylborohydride and sodium triethylborohydride. It can also be utilized as an initiator in radical cyclization reactions.

Application

Reagent for:
  • Enantioselective umpolung allylation of aldehydes
  • Preparation of tetramethylammonium trialkylphenylborate salts

Catalyst for:
  • Radical reductions of alkyl bromides and iodides bearing electron withdrawing groups with N-heterocyclic carbene boranes
  • Synthesis of 1-substituted pyrrolines by N-diallylation of amines and ring-closing metathesis
  • Decarboxylative C-C bond cleavage reactions
  • Alkene hydrogenations
  • Aminyl radical cyclizations onto silyl enol ethers

Modifier for single-site organochromium ethylene polymerization catalysts
Used with lithium tri-tert-butoxyaluminohydride (cat. no. L2904) in the reductive cleavage of ethers and epoxides. Used in the deoxygenation of primary and secondary alcohols.

Packaging

Supplied in a Sure/Pac cylinder and has a 1/4-turn bronze ball valve with a female 1/4" NPT outlet thread (Z122661) installed. Before using the cylinder, ensure that the valve is closed, then remove carbon steel plug that seals the outlet valve.

Compatible with the following:For instructions please refer to technical bulletin AL-136: The Aldrich Sure/Pac Cylinder System

Legal Information

Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Sure/Pac is a trademark of Sigma-Aldrich Co. LLC

also commonly purchased with this product

Product No.
Description
Pricing

septum inlet adapter

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Pyr. Liq. 1 - Skin Corr. 1A

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Convenient Preparation of Trimethylborane and Triethylborane
Koster, R, et al.
Synth. React. Inorg. Met.-Org. Chem. , 3(4), 359-367 (1973)
Masafumi Ueda et al.
Organic letters, 11(20), 4632-4635 (2009-10-09)
Intermolecular hydroxyalkylation of alpha,beta-unsaturated imines involving Et3B-mediated regioselective alkyl radical addition and subsequent hydroxylation with molecular oxygen has been developed, in which N-borylenamine generated by trapping of the enaminyl radical with Et3B was a key intermediate in the proposed aerobic
Tetrahedron, 49, 7193-7193 (1993)
Triethylborane-mediated atom transfer radical cyclization reaction in water
Yorimitsu H, et al.
The Journal of Organic Chemistry, 63(23), 8604-8605 (1998)
Aldrichimica Acta, 6, 21-21 (1973)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service