Skip to Content
Merck
All Photos(1)

Documents

1350400

USP

Isopropyl myristate

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Isopropyl tetradecanoate, Myristic acid isopropyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)12COOCH(CH3)2
CAS Number:
Molecular Weight:
270.45
Beilstein:
1781127
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

isopropyl myristate

manufacturer/tradename

USP

refractive index

n20/D 1.434 (lit.)

bp

193 °C/20 mmHg (lit.)

mp

~3 °C (lit.)

density

0.85 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CCCCCCCCCCCCCC(=O)OC(C)C

InChI

1S/C17H34O2/c1-4-5-6-7-8-9-10-11-12-13-14-15-17(18)19-16(2)3/h16H,4-15H2,1-3H3

InChI key

AXISYYRBXTVTFY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

related product

Storage Class Code

10 - Combustible liquids

WGK

awg

Flash Point(F)

>302.0 °F

Flash Point(C)

> 150 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Hong Du et al.
Drug development and industrial pharmacy, 40(10), 1330-1339 (2013-07-26)
The difference between the chemical polarities of the two categories of active chemical constituents in Chinese angelica volatile oil (CAVO) and Chinese angelica water extract (CAWE) greatly limit the development and clinical application of Chinese angelica preparation. The aim of
Sung Giu Jin et al.
Archives of pharmacal research, 38(2), 216-222 (2014-07-06)
The aim of this research was to evaluate the gel properties, skin permeation and in vivo drug efficacy of a novel dexibuprofen-loaded emulsion gel for topical delivery. In this study, the dexibuprofen-loaded emulsion gel and ibuprofen-loaded emulsion gel were prepared
Jaydeep Patel et al.
Drug delivery, 21(6), 412-435 (2013-11-13)
The present investigation was aimed to develop self-nanoemulsifying tablets (SNETs) as novel nanosized solid oral dosage forms for Irbesartan (IRB). In the first part of the investigation, IRB-loaded self-nanoemulsifying drug delivery systems (SNEDDS) were developed using Capryol 90 - Cremophor
Abdulrahman A Alsultan et al.
Journal of medical microbiology, 63(Pt 11), 1460-1466 (2014-08-12)
Carbapenem-resistant Acinetobacter baumannii (CR-AB) represents a major health-care problem, causing high rates of morbidity and mortality. This study investigated the clonality of CR-AB isolated from diabetic patients from different regions in Saudi Arabia, as well as the relatedness of the
Satomi Asai et al.
Journal of medical microbiology, 63(Pt 11), 1517-1523 (2014-08-22)
A series of clinical isolates of drug-resistant (DR) Acinetobacter baumannii with diverse drug susceptibility was detected from eight patients in the emergency intensive care unit of Tokai University Hospital. The initial isolate was obtained in March 2010 (A. baumannii Tokai

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service