1269254
USP
Famotidine Related Compound E
United States Pharmacopeia (USP) Reference Standard
Synonym(s):
N,N′′′-[Dithiobis(methylene-4,2-thiazolediyl)]bisguanidine, 2,2′-[4,4′-Disulfanediylbis(methylene)bis(thiazole-4,2-diyl)]diguanidine
About This Item
Recommended Products
biological source
synthetic
grade
pharmaceutical primary standard
Agency
USP
API family
famotidine
form
powder
packaging
pkg of 25 mg
manufacturer/tradename
USP
storage condition
protect from light
color
white
application(s)
pharmaceutical (small molecule)
format
neat
storage temp.
2-8°C
SMILES string
[s]1c(nc(c1)CSSCc2nc([s]c2)N=C(N)N)N=C(N)N
InChI
1S/C10H14N8S4/c11-7(12)17-9-15-5(1-19-9)3-21-22-4-6-2-20-10(16-6)18-8(13)14/h1-2H,3-4H2,(H4,11,12,15,17)(H4,13,14,16,18)
InChI key
ZWHJVLVEEDAPHN-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.
Application
Also, for use with USP monograph such as Famotidine
Analysis Note
Other Notes
related product
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service