Skip to Content
Merck
All Photos(1)

Key Documents

PHL89807

Tetrahydropalmatine

phyproof® Reference Substance

Synonym(s):

2,3,9,10-Tetramethoxyberbine, 5,8,13,13a-Tetrahydro-2,3,9,10-tetramethoxy-6H-dibenzo[a,g]quinolizine, Berbine, THP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H25NO4
CAS Number:
Molecular Weight:
355.43
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥98.0% (HPLC)

form

solid

manufacturer/tradename

PhytoLab

SMILES string

N21C(Cc4c(c(c(cc4)OC)OC)C2)c3c(cc(c(c3)OC)OC)CC1

InChI

1S/C21H25NO4/c1-23-18-6-5-13-9-17-15-11-20(25-3)19(24-2)10-14(15)7-8-22(17)12-16(13)21(18)26-4/h5-6,10-11,17H,7-9,12H2,1-4H3

InChI key

AEQDJSLRWYMAQI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by Phytolab GmbH & Co. KG

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service