Skip to Content
Merck
All Photos(1)

Key Documents

PHL89251

Monocrotaline

phyproof® Reference Substance

Synonym(s):

Crotaline, Monocrotaline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H23NO6
CAS Number:
Molecular Weight:
325.36
Beilstein:
48732
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥95.0% (HPLC)

manufacturer/tradename

PhytoLab

mp

204 °C (dec.) (lit.)

application(s)

food and beverages

format

neat

SMILES string

[H][C@@]12[C@H]3CCN1CC=C2COC(=O)[C@](C)(O)[C@](C)(O)[C@@H](C)C(=O)O3

InChI

1S/C16H23NO6/c1-9-13(18)23-11-5-7-17-6-4-10(12(11)17)8-22-14(19)16(3,21)15(9,2)20/h4,9,11-12,20-21H,5-8H2,1-3H3/t9-,11+,12+,15+,16-/m0/s1

InChI key

QVCMHGGNRFRMAD-XFGHUUIASA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Check out our entire range of pyrrolizidine alkaloid standards

Analysis Note

For the analysis of the phytotoxins. Pyrrolizidin alkaloid (PA)

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral - Carc. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Yue Wang et al.
Pharmacology, 105(7-8), 405-415 (2020-01-30)
The regulatory network of aquaporin (AQP) 1 and renin-angiotensin-aldosterone (ALDO) system are not quite clear in pulmonary arterial hypertension (PAH). Thus, we explored the role of AQP1, ALDO and spirolactone (SP) in the PAH animal model and pulmonary arterial smooth

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service