Skip to Content
Merck
All Photos(1)

Key Documents

PHL89243

Linalyl acetate

phyproof® Reference Substance

Synonym(s):

3,7-Dimethyl-1,6-octadien-3-yl acetate, Bergamol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2C(CH=CH2)(CH3)CH2CH2CH=C(CH3)2
CAS Number:
Molecular Weight:
196.29
Beilstein:
1724500
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

primary reference standard

vapor density

6.8 (vs air)

vapor pressure

0.1 mmHg ( 20 °C)

product line

phyproof® Reference Substance

Assay

≥90.0% (GC)

form

liquid

manufacturer/tradename

PhytoLab

refractive index

n20/D 1.453 (lit.)

bp

220 °C (lit.)

density

0.901 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

C\C(C)=C\CCC(C)(OC(C)=O)C=C

InChI

1S/C12H20O2/c1-6-12(5,14-11(4)13)9-7-8-10(2)3/h6,8H,1,7,9H2,2-5H3

InChI key

UWKAYLJWKGQEPM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Other Notes

This compound is commonly found in plants of the genus: salvia thymus

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1B

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

201.2 °F - closed cup

Flash Point(C)

94 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service