Skip to Content
Merck
All Photos(1)

Key Documents

PHL80577

Cyanidin 3-rutinoside chloride

phyproof® Reference Substance

Synonym(s):

Keracyanin chloride, Cyanidin-3-O-rutinoside chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H31ClO15
CAS Number:
Molecular Weight:
630.98
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥90.0% (HPLC)

manufacturer/tradename

PhytoLab

application(s)

food and beverages

format

neat

storage temp.

−20°C

SMILES string

[Cl-].C[C@@H]1O[C@@H](OC[C@H]2O[C@@H](Oc3cc4c(O)cc(O)cc4[o+]c3-c5ccc(O)c(O)c5)[C@H](O)[C@@H](O)[C@@H]2O)[C@H](O)[C@H](O)[C@H]1O

InChI

1S/C27H30O15.ClH/c1-9-19(32)21(34)23(36)26(39-9)38-8-18-20(33)22(35)24(37)27(42-18)41-17-7-12-14(30)5-11(28)6-16(12)40-25(17)10-2-3-13(29)15(31)4-10;/h2-7,9,18-24,26-27,32-37H,8H2,1H3,(H3-,28,29,30,31);1H/t9-,18+,19-,20+,21+,22-,23+,24+,26+,27+;/m0./s1

InChI key

ADZHXBNWNZIHIX-XYGAWYNKSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Application


  • Metabolomic and Transcriptomic Profiling in Plant Sciences: The study explores the impact of Cyanidin 3-rutinoside chloride on color differentiation in Scutellaria baicalensis Georgi. flowers, utilizing metabolomic and transcriptomic profiling to uncover the biochemical pathways influenced by this compound, highlighting its role in anthocyanin research and its potential antioxidant activities (Wang et al., 2022).

  • Optimization of Anthocyanin Extraction: Research on the extraction of anthocyanins from Borage (Echium amoenum) flowers using environmentally friendly solvents demonstrates Cyanidin 3-rutinoside chloride′s antioxidant activity and its bioavailability, positioning it as a crucial flavonoid compound for studies in antioxidant biochemistry and natural dye applications in pharmaceutical research (Zannou et al., 2021).

Other Notes

This compound is commonly found in plants of the genus: sambucus

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service