Skip to Content
Merck
All Photos(1)

Key Documents

805106

TSKgel® G3000PW (12 µm) HPLC Columns

L x I.D. 60 cm × 7.5 mm, hydroxylated methacrylate phase

Synonym(s):

TSKgel® Size Exclusion (PW-Type) HPLC Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711
eCl@ss:
32110501

product name

TSKgel® G3000PW HPLC Column, phase hydroxylated methacrylate, L × I.D. 60 cm × 7.5 mm, 12 μm particle size

material

stainless steel hardware

Agency

suitable for USP L37
suitable for USP L38
suitable for USP L39

product line

TSKgel®

manufacturer/tradename

Tosoh 05106

parameter

≤0.5 M salt concentration
≤20% organic solvent
1.2 mL/min flow rate
40 bar max. pressure
80 °C temperature

technique(s)

HPLC: suitable

L × I.D.

60 cm × 7.5 mm

matrix

polymer particle platform

matrix active group

hydroxylated methacrylate phase

particle size

12 μm

pore size

200 Å

operating pH

2-12

separation technique

size exclusion (SEC)

Looking for similar products? Visit Product Comparison Guide

General description

Use the TSKgel G3000PW column for the analysis of water-soluble polymers of less than 50,000 Da molecular weight.

Physical form

Shipped in water.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service