Skip to Content
Merck
All Photos(1)

Key Documents

805103

TSKgel® G3000SW (10 µm) HPLC Columns

L x I.D. 60 cm × 7.5 mm, diol phase

Synonym(s):

TSKgel® Size Exclusion (SW-Type) HPLC Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
Pricing and availability is not currently available.

Product Name

TSKgel® G3000SW HPLC Column, phase diol, L × I.D. 60 cm × 7.5 mm, 10 μm particle size

material

stainless steel

Agency

suitable for USP L20
suitable for USP L59

product line

TSKgel®

manufacturer/tradename

Tosoh 05103

parameter

≤0.5 M salt concentration
0-100% aqueous soluble organic solvents
1.2 mL/min flow rate
10-30 °C temperature
50 bar max. pressure

technique(s)

HPLC: suitable

L × I.D.

60 cm × 7.5 mm

matrix

silica particle platform

matrix active group

diol phase

particle size

10 μm

pore size

250 Å

operating pH

2.5-7.5

separation technique

size exclusion (SEC)

Looking for similar products? Visit Product Comparison Guide

General description

As the first generation of silica-based SEC columns, TSKgel G3000SW columns provide excellent separation of proteins and other biomolecules with molecular weights up to 500,000 Da. TSKgel G3000SW columns are available in 10μm particle size in Glass and Stainless Steel analytical columns. Particle sizes of 13 are used to pack 30cm and 60cm x 21.5mm ID columns, respectively, for semi-preparative purifications.

Physical form

Shipped in 0.05% NaN3 and 0.1 mol/L Na2SO4 in 0.1 M phosphate buffer, pH 6.7.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Is this column compatible with an Aekta FLPC system? Or does it only run on HPLCs? Thanks!

    1 answer
    1. The operating flow rate and pressures of the column are within the capabilities of the instrument. It is recommended to check with the vendor of the instrument to determine if the specific instrument is set up with the correct connectors and if it is suitably configured to run the intended application.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service