Key Documents
57267
Supelco
Cartridge Adapter for H300 Cartridges
pkg of 6 ea, for use with 12, 20, & 60 mL SPE tubes
About This Item
UNSPSC Code:
12000000
Pricing and availability is not currently available.
packaging
pkg of 6 ea
technique(s)
solid phase extraction (SPE): suitable
application(s)
environmental
food and beverages
compatibility
for use with 12, 20, & 60 mL SPE tubes
General description
Cartridge adapter is generally required for sampling of solid phase extract during SPE.
Choose from one of the most recent versions:
Certificates of Analysis (COA)