Skip to Content
Merck
All Photos(1)

Key Documents

44594-U

Supelco

P-I-A-N-O Kit

analytical standard

Synonym(s):

Piano Assay Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Agency

ASTM® D6729,D6730,D6733

packaging

pkg of 1 kit

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

petroleum

format

multi-component solution

storage temp.

2-8°C

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials

Kit Components Only

Product No.
Description

  • n-Paraffins Mix (44585-U) .1 mL

  • Isoparaffins Mix (44586-U) .1 mL

  • Aromatics Mix (44587) .1 mL

  • Naphthenes Mix (44588) .1 mL

  • Olefins Mix (44589) .1 mL

  • P-I-A-N-O Mix (44593-U) .1 mL

analytical column

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Carc. 1A - Eye Dam. 1 - Flam. Liq. 2 - Muta. 1B - Repr. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1 Inhalation - STOT RE 2 - STOT SE 3

Target Organs

Blood, Central nervous system, Nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids

Flash Point(F)

-57.0 °F

Flash Point(C)

-49.45 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Complete offer for petrochemical testing including environmental and industrial hygiene monitoring solutions.

Complete offer for petrochemical testing including environmental and industrial hygiene monitoring solutions.

Complete offer for petrochemical testing including environmental and industrial hygiene monitoring solutions.

Complete offer for petrochemical testing including environmental and industrial hygiene monitoring solutions.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service