Skip to Content
Merck
All Photos(1)

Key Documents

442499

Supelco

Bromodichloroacetic acid

analytical standard

Synonym(s):

Bromdichloroacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrCl2CCOOH
CAS Number:
Molecular Weight:
207.84
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

Quality Level

CofA

current certificate can be downloaded

packaging

ampule of 100 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

69-72 °C (lit.)

application(s)

environmental

format

neat

storage temp.

2-30°C

SMILES string

OC(=O)C(Cl)(Cl)Br

InChI

1S/C2HBrCl2O2/c3-2(4,5)1(6)7/h(H,6,7)

InChI key

XSWVFEQKZFUULO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Bromodichloroacetic acid belongs to the class of haloacetic acids (HAAs) which are organic disinfection byproducts produced in chlorinated water. They are common contaminants observed in drinking water.
Sigma-Aldrich offers bromodichloroacetic acid for a variety of applications that include, but are not limited to, environmental, food and beverage, forensics, petrochemical and pharmaceutical analysis.

Application

Bromodichloroacetic acid may be used as an analytical reference standard for the determination of the analyte in environmental samples by various chromatographic samples.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Rapid IC-ICP/MS method for simultaneous analysis of iodoacetic acids, bromoacetic acids, bromate, and other related halogenated compounds in water
Shi H and Adams C
Talanta, 79(2), 523-527 (2009)
The application of gas chromatography to environmental analysis
Santos FJ and Galceran MT
TrAC, Trends in Analytical Chemistry, 21(9-10), 672-685 (2002)
E W Austin et al.
Journal of toxicology and environmental health, 52(4), 367-383 (1997-11-14)
Haloacetates are a common class of water chlorination by-products. Depending on the amount of bromide in the source water, varying amounts of chlorinated, brominated, and mixed bromochloro haloacetates are produced. When administered to rodents, haloacetates have been shown to increase
J L Merdink et al.
Journal of applied toxicology : JAT, 21(1), 53-57 (2001-02-17)
The oral and i.v. elimination kinetics were investigated for bromodichloroacetate (BDCA), a haloacetate found in drinking water. The BDCA was administered at a dose of 5, 20 and 100 mg kg-1 to B6C3F1 mice and appears to distribute to the
G Xu et al.
Drug metabolism and disposition: the biological fate of chemicals, 23(12), 1412-1416 (1995-12-01)
Trichloroacetate (TCA), dichloroacetate (DCA), and bromodichloroacetate (BDCA) are byproducts of the chlorination of drinking water. TCA acts primarily as a peroxisome proliferator, but DCA produces tumors at doses less than required for peroxisome proliferation. BDCA does not induce peroxisome proliferation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service