Key Documents
33183
Supelco
Hand Decapper
for use with 20mm crimp seals, pkg of 1 ea
About This Item
UNSPSC Code:
41122001
Pricing and availability is not currently available.
description
Hand decapper
packaging
pkg of 1 ea
compatibility
for use with 20mm crimp seals
General description
Hand decapper is used for quick and easy removal of a 20mm aluminum seal.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Articles
Learn more about Biodiesel as a renewable, alternative diesel fuel produced from vegetable oils, animal fats, or recycled restaurant grease.
Learn more about Biodiesel as a renewable, alternative diesel fuel produced from vegetable oils, animal fats, or recycled restaurant grease.
Learn more about Biodiesel as a renewable, alternative diesel fuel produced from vegetable oils, animal fats, or recycled restaurant grease.
Learn more about Biodiesel as a renewable, alternative diesel fuel produced from vegetable oils, animal fats, or recycled restaurant grease.