22992
Oxiclear™ Disposable Oxygen/Moisture Trap
1/8 in. fitting
Synonym(s):
DISPOSABLE PURIFIER 1/8 EA
About This Item
Recommended Products
fitting
1/8 in. fitting
Looking for similar products? Visit Product Comparison Guide
General description
Dimensions:
- Oxiclear: 7 1/2 in. (19.0 cm) long x 1 7/8 in. (48 mm) diameter
Legal Information
Storage Class Code
13 - Non Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service