Key Documents
20977
Hamilton® GASTIGHT® syringe, 1700 series, luer tip
1702LT, volume 25 μL, without needle, Luer Tip
About This Item
UNSPSC Code:
41122001
material
glass syringe
needle size
26 ga
feature
without needle Luer Tip
packaging
pkg of 1 ea
manufacturer/tradename
Hamilton 80201
volume
25 μL
General description
Hamilton syringe is available in gas-tight type that minimizes the gas exchange in GC analysis. It gives accurate volume delivery. The plunger is mostly unsurpassed. The tip is threaded in a luer lock syringe allowing the needle to be tightly screwed to the tip.
Application
It was used to inject nutritional supplement the study animals (rats & mice) to understand choroidal neovascularization development and reduction of acute conjunctivitis inflammation.
Legal Information
GASTIGHT is a registered trademark of Hamilton Co.
Hamilton is a registered trademark of Hamilton Co.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Azithromycin reduces inflammation in a rat model of acute conjunctivitis.
Fernandez-Robredo, Patricia, et al.
Molecular Vision, 155-165 (2013)
Modifying choroidal neovascularization development with a nutritional supplement in mice.
Ivanescu, Alina Adriana, et al.
Nutrients, 19, 153-165 (2013)
Guy, Jeffrey S.
Pharmacology for the Prehospital Professional, 54-54 (2009)
Wolf, Don P.
In Vitro Fertilization and Embryo Transfer: A Manual of Basic Techniques, 06-Dec, 192-192 (2012)
Wolf, Don P.
In Vitro Fertilization and Embryo Transfer: A Manual of Basic Techniques, 06-Dec, 192-192 (2012)