Skip to Content
Merck
All Photos(1)

Key Documents

20011

Supelco

Succinylacetone

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

4,6-Dioxoheptanoic acid, Succinylacetone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCH2COCH2CH2CO2H
CAS Number:
Molecular Weight:
158.15
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

mp

66-67 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(=O)CC(=O)CCC(O)=O

InChI

1S/C7H10O4/c1-5(8)4-6(9)2-3-7(10)11/h2-4H2,1H3,(H,10,11)

InChI key

WYEPBHZLDUPIOD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to the SI through a primary reference material from an NMI, e.g. NIST, NRC or NMIJ.
Certified content incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com

Biochem/physiol Actions

An inhibitor of heme biosynthesis

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service