Skip to Content
Merck
All Photos(1)

Documents

13629-U

Supelco

MCI GEL® CHP20P Polymeric Adsorbent

700 mesh, bottle of 100g

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151817
NACRES:
SB.52

product name

MCI GEL® CHP20P, bottle of 100 g

form

solid

packaging

bottle of 100 g

technique(s)

LPLC: suitable
solid phase extraction (SPE): suitable

surface area

~500 m2/g

matrix

styrene-divinylbenzene

matrix active group

polymer

particle size

75-150 μm

pore size

~1.30 mL/g pore volume
400-600 Å mean pore size

density

1.01 g/mL at 25 °C (true wet)(lit.)

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

MCI GEL® CHP20P is a highly porous styrene-divinylbenzene polymer resin used as column packing material.

Application

MCI GEL® CHP20P was used in testing solid phase enrichment of phosphomolybdenum blue.
Polyaromatic adsorbent resin designed for biopharmaceutical separations: aromatic compounds, peptides, steroids, desalting, etc. Very good for reversed phase chromatography applications, even with nonaqueous solvents.

Legal Information

MCI GEL is a registered trademark of Mitsubishi Chemical Corp.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

N Okamura et al.
Phytochemistry, 37(5), 1463-1466 (1994-11-01)
Three new flavonoid glucuronides, luteolin 3'-O-beta-D-glucuronide, luteolin 3'-O-(4"-O-acetyl)-beta-D-glucuronide, and luteolin 3'-O-(3"-O-acetyl)-beta-D-glucuronide, together with hesperidin, were isolated from 50% aqueous MeOH extract of the leaves of rosemary. The structures were established by chemical and spectroscopic methods. Their antioxidant activities were evaluated
Highly sensitive flow analysis determination of orthophosphate using solid phase enrichment of phosphomolybdenum blue without need for organic solvents in elution.
Heckemann, H-J.
Analytica Chimica Acta, 410, 177-184 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service