Skip to Content
Merck
All Photos(1)

Key Documents

12100AST

Supelco

Astec® CHIROBIOTIC® T Chiral (5 μm) HPLC Columns

L × I.D. 2 cm × 4 mm, HPLC Guard Cartridge

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
SB.52
Pricing and availability is not currently available.

Product Name

Astec® CHIROBIOTIC® T Chiral HPLC Guard Column, Cartridge, 5 μm particle size, L × I.D. 2 cm × 4 mm

material

stainless steel column

Agency

suitable for USP L63

product line

Astec®

packaging

pkg of 1 ea

manufacturer/tradename

Astec®

parameter

0-45 °C temperature
241 bar pressure (3500 psi)

technique(s)

HPLC: suitable
LC/MS: suitable

L × I.D.

2 cm × 4 mm

matrix

fully porous particle

matrix active group

teicoplanin glycopeptide phase

particle size

5 μm

pore size

100 Å

operating pH

3.8-6.8

separation technique

chiral

Looking for similar products? Visit Product Comparison Guide

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

László Sipos et al.
Journal of chromatography. A, 1217(44), 6956-6963 (2010-09-25)
The enantiomers of five monoterpene-based 2-amino carboxylic acids were directly separated on chiral stationary phases containing macrocyclic glycopeptide antibiotics such as teicoplanin (Astec Chirobiotic T and T2) and teicoplanin aglycone (Chirobiotic TAG) as chiral selectors. The effects of pH, the
Xingxing Diao et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 939, 67-72 (2013-10-10)
A novel and sensitive liquid chromatography-tandem mass spectrometry (LC-MS/MS) method was developed and validated to determine the exposure of 3-n-butylphthalide (NBP) enantiomers in human plasma. The NBP enantiomers were extracted from human plasma using methyl tert-butyl ether. The baseline separation
Jolanta Flieger et al.
Journal of separation science, 40(11), 2374-2381 (2017-04-21)
We present the specific cooperative effect of a semisynthetic glycopeptide antibiotic teicoplanin and chiral ionic liquids containing the (1R,2S,5R)-(-)-menthol moiety on the chiral recognition of enantiomers of mandelic acid, vanilmandelic acid, and phenyllactic acid. Experiments were performed chromatographically on an
László Sipos et al.
Journal of chromatography. A, 1232, 142-151 (2011-12-20)
The enantiomers of four unusual isoxazoline-fused 2-aminocyclopentanecarboxylic acids were directly separated on chiral stationary phases containing macrocyclic glycopeptide antibiotics teicoplanin (Astec Chirobiotic T and T2), teicoplanin aglycone (Chirobiotic TAG), vancomycin (Chirobiotic V) and vancomycin aglycone (Chirobiotic VAG) as chiral selectors.
Róbert Berkecz et al.
Journal of chromatography. A, 1216(6), 927-932 (2008-12-27)
The direct separation of the enantiomers of four 2-aminomono- or dihydroxycyclopentanecarboxylic acids and four 2-aminodihydroxycyclohexanecarboxylic acids was performed on chiral stationary phases containing macrocyclic glycopeptide antibiotics such as teicoplanin (Astec Chirobiotic T and T2), teicoplanin aglycone (Chirobiotic TAG) or ristocetin

Questions

  1. What is the best chiral guard column for Astec® CHIROBIOTIC® T Chiral HPLC Column 250 x 10 mm (part number 12034AST and/or 14034AST)?

    1 answer
    1. Guard columns for the Astec Chirobiotic columns are available in 1 mm (11101AST) and 4 mm (11100AST) IDs. For the 10 mm ID Astec columns,it is recommend using the 4 mm ID guard. Although it is only a 4 mm guard column, it may need to adjust the flow rate and monitor backpressure. This solution is the best option as 10 mm guard is longer available.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service