Skip to Content
Merck
All Photos(20)

Key Documents

ZRB001906

Sigma-Aldrich

Anti-ATRX Antibody, clone 2A12, ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

ATP-dependent helicase ATRX, EC: 3.6.4.12, Transcriptional regulator ATRX, X-linked helicase II, X-linked nuclear protein, XNP, Znf-HX

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit (recombinant)

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

2A12, recombinant monoclonal
monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 282.58 kDa
observed mol wt ~280 kDa

species reactivity

mouse, human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

immunocytochemistry: suitable using 1:100
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable using 1:100
western blot: 1:1,000-1:4,000

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.


Learn more about ZooMAb here.

Specificity

Clone 2A12 is a ZooMAb rabbit recombinant monoclonal antibody that detects Transcriptional regulator ATRX. It targets an epitope with in 141 amino acids from the C-terminal region.

Immunogen

GST/His-tagged recombinant fragment corresponding to 141 amino acids from the C-terminal region of human Transcriptional regulator ATRX (isoform 4).

Application

Anti-ATRX, clone 2A12, ZooMAb, Cat. No. ZRB001906, is a recombinant Rabbit Monoclonal Antibody that specifically detects ATRX and has been tested for use in Immunocytochemistry, Immunohistochemistry (Paraffin), and Western Blotting.
Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected ATRX in human prostate tissue sections.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected ATRX in HeLa, A431, HUVEC, and NIH 3T3 cells.

Western Blot Analysis: A 1:1,000 - 1:4,000 dilution from a representative lot.

Target description

Transcriptional regulator ATRX (UniProt: P46100; also known as EC:3.6.4.12, ATP-dependent helicase ATRX, X-linked helicase II, X-linked nuclear protein, XNP, Znf-HX, ATRX) is encoded by the ATRX (also known as RAD54L, XH2) gene (Gene ID: 546) in human. ATRX is ubiquitously distributed and is involved in transcriptional regulation and chromatin remodeling. ATRX has a PHD zinc finger motif and an ATPase/helicase domain of the SWI/SNF type. It facilitates DNA replication in multiple cellular environments and is required for efficient replication of a subset of genomic loci. ATRX binds to DNA tandem repeat sequences in both telomeres and euchromatin and is shown to bind quadruplex structures in vitro. ATRX associates with pericentromeric heterochromatin during interphase and mitosis, probably by interacting with CBX5/HP1 alpha. It interacts with DAXX to form the chromatin remodeling complex ATRX:DAXX, which has ATP-dependent DNA translocase activity and catalyzes the replication-independent deposition of histone H3.3 in pericentric DNA repeats outside S-phase and telomeres. ATRX is believed to be involved in telomere maintenance via recombination in ALT (alternative lengthening of telomeres) cell lines. ATRX is phosphorylated at serine residues during mitosis and phosphorylation is most evident at pro-metaphase stage. Phosphorylation of ATRX is necessary for release of the protein from the nuclear matrix and that it may facilitate progression to mitosis. Dephosphorylation of ATRX seems to coincide with exit of the cells from M phase. Mutations in ATRX gene are known to cause severe psychomotor and mental retardation, facial dysmorphism, and urogenital abnormalities. This ZooMAb recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Berube, NG et al. (2000). Hum. Mol. Gen. 9(4):539-547).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

Reconstitute lyophilized antibody pellet with 25μL of ultrapure water or Phosphate Buffered Saline (PBS). Please refer to our reconstitution protocol and the specific application guidance on the suggested starting dilutions and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service