Skip to Content
Merck
All Photos(1)

Key Documents

Z708542

Sigma-Aldrich

Square weighing boats

size S

Synonym(s):

weigh boat, weigh dish, weighing dish, weighing pan

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121812
NACRES:
NB.71
Pricing and availability is not currently available.

material

polystyrene
square

size

S

packaging

pack of 500 ea

manufacturer/tradename

Heathrow Scientific HS1421A

L × W × H

41 mm × 41 mm × 8 mm

color

blue

Looking for similar products? Visit Product Comparison Guide

General description

Polystyrene weighing boats with a smooth surface to provide accurate pour-outs with minimal sample loss. Shallow shape with a wide, flat bottom to resist tipping. Rounded corners simplify transfer. Biologically inert and resistant to dilute and weak acids, aqueous solutions, alcohols and bases. May be used with microwave moisture analyzers. Resist temperatures to 200 °F (93 °C).

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service