Z707805
TPP® tissue culture plates
24 well plate, flat bottom, polystyrene, 1.86 cm2, sterile, 72/cs
Synonym(s):
24 well plate, cell culture plate, tissue culture plate
About This Item
flat bottom clear polystyrene wells
polystyrene
Recommended Products
material
flat bottom (F-base)
flat bottom clear polystyrene wells
polystyrene
sterility
sterile
packaging
case of 72
manufacturer/tradename
TPP 92424
availability
not available in (available only in North America)
external L × W × H
128 mm × 85 mm × 22 mm
size
24 wells
surface area
1.86 cm2 , growth surface (cm2)
working volume
6.3 mL
suitability
suitable for (cell culture; tissue culture applications)
binding type
Tissue Culture (TC)-treated surface
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Advantages of the TPP tissue culture plates:
- Lid with air-venting system guarantees controlled gas exchange with low evaporation
- Sloped edge allows placement of the lid in one position only
- Yellow marking area on lid and plate ensure correct lid orientation: yellow on yellow – match!
- Ridged grip area ensures a secure grasp and prevents from accidentally lifting off the lid
- Clear alpha-numerical labelling between the wells for easy orientation during microscopy, easy to see black alpha-numerics on the side
- Growth area on spherical zone only
- Absolutely flat growth surface
- All TPP plate sizes (6, 12, 24 and 96-well) stack securely together
- Air vents in plate base prevent condensation and provide consistent air-flow between stacked plates
- Crystal clear transparency
Advantages of the TPP 96 well tissue culture plates:
- Manufactured according to ANSI/SBS-3d-standards
- Standardized with an interleaved barcode 2/5
- White base colour is embossed firmly on the test plate; loss of identification through detachment is excluded
- 2 well geometries available: F-base and U-base
Well geometry for 96 well tissue culture plates
F-base (flat bottom):
- Excellent optical characteristics
- Suitable for precise optical measurements the measuring light is not distracted by the geometry-as well as microscopy applications
U-base (round bottom):
- No rims well suited for pipetting
- Used for agglutination tests and „+/-„ evaluations
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service