Gas exchange studies of this sealing film have not been performed. However, based on the polyethylene composition, these films are not expected to be permeable.
Z688630
Titer Tops® sealing film for microplates
pk of 100 ×
Synonym(s):
microplate sealing film, multiwell plate sealing film, sealing film
About This Item
Recommended Products
material
film (polyurethane)
polyethylene
sterility
sterile
feature
piercable
prevents well-to-well contamination, sample spillage and evaporation
manufacturer/tradename
Diversified Biotech T-TOPS-100
packaging
pk of 100 ×
W × L
8.26 cm × 15.24 cm
color
clear
General description
- Durable and thick (4.4 mil) yet easily pierced with a pipette tip
- Prevents well-to-well contamination, sample spillage and evaporation
- Polyethylene film that provides excellent adhesion to plastic microplates
- Non-cytotoxic, sterile, solvent resistant and waterproof
- Stays solidly in place even when exposed to 100% humidity and extreme temperatures ranging from -40 °C to +100 °C (Not recommended for use in thermal cyclers)
- Chemically resistant and thermally stable
- Easily marked with any writing instrument
- Two scored end taps for easy positioning
- UV transparent down to 250 nm
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
Are these seals permeable for gusses?
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service