Z326704
BRAND® BLAUBRAND® ETERNA volumetric flask
volume 5 mL, accuracy: 0.04 mL
All Photos(1)
About This Item
Recommended Products
material
borosilicate glass
wide-neck
Joint
joint ST/NS neck10/19
joint ST/NS10/19
description
calibrated to contain (TC, EX)
sterility
non-sterile
feature
accuracy class A (DIN EN ISO 1042, calibrated to contain (TC, EX))
closure type PP Stopper
packaging
pack of 2
manufacturer/tradename
BRAND
volume measuring range
5 mL, accuracy: 0.04 mL
capacity
5 mL
General description
Class A, conformity certified. Made from Boro 3.3, Supplied with a square grip PP stopper. Marks and inscriptions in ETERNA amber diffusion stain, particularly resistant to aggressive cleaning methods. TO DIN EN ISO 1042.
All BLAUBRAND® volumetric flasks are supplied with a batch number and an accompanying batch certificate in the original packaging.
All BLAUBRAND® volumetric flasks are supplied with a batch number and an accompanying batch certificate in the original packaging.
Other Notes
BR36838 (India)
Legal Information
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG