V2502
Marine venom from Chrysaora quinquecirrha (Sea Nettle)
lyophilized powder
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352200
NACRES:
NA.77
Recommended Products
form
lyophilized powder
storage temp.
−20°C
Biochem/physiol Actions
Sea nettle, Chrysaora quinquecirrha venom (CQV), is toxic to the cardiac, respiratory, renal, and hepatic systems in animal models.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service