Skip to Content
Merck
All Photos(1)

Key Documents

U4133

Sigma-Aldrich

URB597

≥98% (HPLC), powder

Synonym(s):

Cyclohexylcarbamic acid 3´-carbamoyl-biphenyl-3-yl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H22N2O3
CAS Number:
Molecular Weight:
338.40
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white

solubility

DMSO: soluble ~14 mg/mL

storage temp.

2-8°C

SMILES string

NC(=O)c1cccc(c1)-c2cccc(OC(=O)NC3CCCCC3)c2

InChI

1S/C20H22N2O3/c21-19(23)16-8-4-6-14(12-16)15-7-5-11-18(13-15)25-20(24)22-17-9-2-1-3-10-17/h4-8,11-13,17H,1-3,9-10H2,(H2,21,23)(H,22,24)

InChI key

ROFVXGGUISEHAM-UHFFFAOYSA-N

General description

URB597 binds to active sites of fatty acid amide hydrolases and inhibits their activity. URB597 alters expression of tyrosine hydroxylase and interacts with abnormal-cannabidiol (Abn-CBD) and peroxisome proliferator-activated receptors (PPARs). URB597 elicits antinociception property via cannabinoid receptor by maintaining endocannabinoid anandamide (AEA) levels. URB597 reduces abnormal hyperactivity in neurons and could be for treatment of seizures and improving synaptic plasticity.

Biochem/physiol Actions

Potent, selective fatty acid amide hydrolase (FAAH) inhibitor.

Preparation Note

URB597 is soluble in DMSO at a concentration that is approximately 14 mg/ml.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lucas Albrechet-Souza et al.
Neurobiology of stress, 15, 100387-100387 (2021-09-16)
Understanding sex differences in behavioral and molecular effects of stress has important implications for understanding the vulnerability to chronic psychiatric disorders associated with stress response circuitry. The amygdala is critical for emotional learning and generating behavioral responses to stressful stimuli
Luara A Batista et al.
Behavioural brain research, 317, 508-514 (2016-10-28)
Selective stimulation of carotid chemoreceptors by intravenous infusion of low doses of potassium cyanide (KCN) produces short-lasting escape responses that have been proposed as a model of panic attack. In turn, preclinical studies suggest that facilitation of the endocannabinoid system
Interaction between the cholecystokinin and endogenous cannabinoid systems in cued fear expression and extinction retention
Bowers ME and Ressler KJ
Neuropsychopharmacology, 30, 688-688 (2015)
The FAAH inhibitor URB597 suppresses hippocampal maximal dentate afterdischarges and restores seizure-induced impairment of short and long-term synaptic plasticity
Colangeli R, et al.
Scientific Reports, 7(1), 11152-11152 (2017)
The FAAH inhibitor URB597 efficiently reduces tyrosine hydroxylase expression through CB1-and FAAH-independent mechanisms
Bosier B, et al.
British Journal of Pharmacology, 169(4), 794-807 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service