Skip to Content
Merck
All Photos(1)

Key Documents

U3383

Sigma-Aldrich

Urease Buffer Reagent

Synonym(s):

Urea colorimetric determination assay reagent

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
NACRES:
NA.54
Pricing and availability is not currently available.

description

When reconstituted, product is a solution of urease from Jack bean in 27 mM EDTA with 0.1% sodium azide as a preservative.

Quality Level

form

powder

mol wt

545 kDa

storage temp.

2-8°C

General description

Urease is a 545 kDa protein critical for the hydrolysis of urea to carbon dioxide and ammonia. [1]
Urease is a dinickel enzyme that hydrolyses urea into ammonia and carbon dioxide.[2][3] It is secreted by several bacterial species, invertebrates, fungi, and plants.[3] Detection of urease activity is a potential diagnostic tool for Helicobacter pylori infections and several oral bacterial infections.[2] two Ni2+ ions bridged by a hydroxyl group and a carbamylated lysine.[3]

Application

Urease Buffer Reagent has been used:
  • to measure plasma urea N (PUN) by the hypochlorite-phenol nitroprusside method[4]
  • in cell viability assay to measure the synthesis of urea in human fetal stem/progenitor cells[5]
  • to study the metabolic capacity of rat hepatocytes[6]

Used in conjunction with Phenol Nitroprusside solution (Product Code P 6994) and Alkaline Hydpochlorite solution (Product Code A 1727) for the colorimetric determination of urea.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kang Cheng et al.
Hepatology (Baltimore, Md.), 50(4), 1194-1203 (2009-07-29)
Tracking stem/progenitor cells through noninvasive imaging is a helpful means of assessing the targeting of transplanted cells to specific organs. We performed in vitro and in vivo studies wherein adult human hepatocytes and human fetal liver stem/progenitor cells were labeled
Yuta Enami et al.
Hepatology (Baltimore, Md.), 55(4), 1182-1192 (2011-10-14)
Organs from non-heart-beating donors are attractive for use in cell therapy. Understanding the nature of molecular perturbations following reperfusion/reoxygenation will be highly significant for non-heart-beating donor cells. We studied non-heart-beating donor rats for global gene expression with Affymetrix microarrays, hepatic
Cristian Follmer
Phytochemistry, 69(1), 18-28 (2007-08-21)
The broad distribution of ureases in leguminous seeds, as well as the accumulation pattern of the protein during seed maturation, are suggestive of an important physiological role for this enzyme. Since the isolation and characterization of jack bean urease by
L A Winkelman et al.
Journal of dairy science, 96(12), 7565-7577 (2013-10-15)
This study investigated the effect of 2 different types of long-acting insulin on milk production, milk composition, and metabolism in lactating dairy cows. Multiparous cows (n=30) averaging 88 d in milk were assigned to one of 3 treatments in a
Gunnar Dahlén et al.
BMC oral health, 18(1), 89-89 (2018-05-20)
Urease is an enzyme produced by plaque bacteria hydrolysing urea from saliva and gingival exudate into ammonia in order to regulate the pH in the dental biofilm. The aim of this study was to assess the urease activity among oral

Questions

1–3 of 3 Questions  
  1. What's the concentration of Jack bean Urease? I can only see the concentration of buffer.

    1 answer
    1. As provided, this product contains 100 units per vial. The mass weight per vial is not determined.

      Helpful?

  2. How many urease units does 1 vial contain?

    1 answer
    1. As provided, this product contains 100 units per vial.

      Helpful?

  3. What is the volume of water used for reconstitution of 1 vial?

    1 answer
    1. As mentioned on the sample Certificate of Analysis, 30 mL of purified water in one vial of this material yields a clear solution.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service