Skip to Content
Merck
All Photos(1)

Key Documents

TRUEPEG

Sigma-Aldrich

TrueGel3D Crosslinker

PEG non cell-degradable crosslinker

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.71
Pricing and availability is not currently available.

technique(s)

tissue processing: suitable

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

PEG-Link is comprised of polyethylene glycol and thiol (SH) groups at each end. The thiol groups form stable thioether bond with thiol-reactive groups of the polymers (Mal-PVA, Mal-Dextran, SG-PVA and SG-Dextran), which results in stable hydrogel. The matrix metalloproteases secreted by cells cannot cleave PEG crosslinker.

Application

TRUEPEG is used to crosslink PVA and dextran polymers to generate stable hydrogel.

Features and Benefits

Non-MMP degradable.

Components

TRUEPEG contains PEG non cell-degradable crosslinker lyophilized and water.

Caution

Avoid prolong exposure of PEG non cell-degradable crosslinker to air.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Troubleshoot TrueGel3D™ hydrogel formation and cell viability issues with RGD-degradable polymer and CD cell-degradable crosslinker.

Troubleshoot TrueGel3D™ hydrogel formation and cell viability issues with RGD-degradable polymer and CD cell-degradable crosslinker.

Protocols

TrueGel3D hydrogel enables cell encapsulation for 3D culture, mimicking native tissue environment with chemically defined properties.

TrueGel3D hydrogel enables cell encapsulation for 3D culture, mimicking native tissue environment with chemically defined properties.

TrueGel3D™ is a chemically defined hydrogel for 3D cell growth, facilitating microchannel filling with slow gelation.

TrueGel3D hydrogel enables cell encapsulation for 3D culture, mimicking native tissue environment with chemically defined properties.

See All

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service