Skip to Content
Merck
All Photos(1)

Key Documents

SML4058

Sigma-Aldrich

Florfenicol

≥98% (HPLC)

Synonym(s):

(-)-Florfenicol, 2,2-Dichloro-N-((1R,2S)-3-fluoro-1-hydroxy-1-(4-(methylsulfonyl)phenyl)propan-2-yl)acetamide, 2,2-Dichloro-N-[(1S,2R)-1-(fluoromethyl)-2-hydroxy-2-[4-(methylsulfonyl)phenyl]ethyl]acetamide, SCH 25298

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H14Cl2FNO4S
CAS Number:
Molecular Weight:
358.21
MDL number:
UNSPSC Code:
12352200
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

ClC(C(N[C@@H]([C@H](O)C1=CC=C(S(C)(=O)=O)C=C1)CF)=O)Cl

InChI

1S/C12H14Cl2FNO4S/c1-21(19,20)8-4-2-7(3-5-8)10(17)9(6-15)16-12(18)11(13)14/h2-5,9-11,17H,6H2,1H3,(H,16,18)/t9-,10-/m1/s1

InChI key

AYIRNRDRBQJXIF-NXEZZACHSA-N

Biochem/physiol Actions

Broad-spectrum antibiotic that inhibits protein synthesis by binding to the 50S ribosomal subunit of bacteria.



Florfenicol is a broad-spectrum antibiotic that is primarily used in veterinary medicine to treat respiratory infections in cattle and swine, and to prevent Salmonella infection in poultry. It has favorable ADME properties and is a fluorinated analog of chloramphenicol that has good tissue penetration and is resistant to chloramphenicol acetyltransferase inactivation. Florfenicol inhibits ribosomal activity and effectively inhibits bacterial protein synthesis, which makes it effective against various bacteria. However, oral administration of florfenicol may alter animal microbiota.

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 2 - STOT RE 1

Target Organs

Liver,Brain,Testes,Spinal cord,Blood,gallbladder

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xingying Guo et al.
Environmental research, 244, 117934-117934 (2023-12-19)
Florfenicol, as a replacement for chloramphenicol, can tightly bind to the A site of the 23S rRNA in the 50S subunit of the 70S ribosome, thereby inhibiting protein synthesis and bacterial proliferation. Due to the widespread use in aquaculture and
P S Gaunt et al.
Journal of veterinary pharmacology and therapeutics, 36(5), 502-506 (2012-08-14)
Plasma disposition of florfenicol in channel catfish was investigated after an oral multidose (10 mg/kg for 10 days) administration in freshwater at water temperatures ranging from 24.7 to 25.9 °C. Florfenicol concentrations in plasma were analyzed by means of liquid
V P Syriopoulou et al.
Antimicrobial agents and chemotherapy, 19(2), 294-297 (1981-02-01)
We evaluated the in vitro antimicrobial activity of Sch 24893, Sch 25298, and Sch 25393, three novel analogs of chloramphenicol and thiamphenicol. All of the analogs had minimal inhibitory concentrations of less than or equal to 10 micrograms/ml for 18

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service