Skip to Content
Merck
All Photos(1)

Key Documents

SML3650

Sigma-Aldrich

CTPI-2

≥98% (HPLC)

Synonym(s):

2-(4-Chloro-3-nitrobenzenesulfonamido)benzoic acid, 2-[[(4-Chloro-3-nitrophenyl)sulfonyl]amino]benzoic acid, CTPI 2, CTPI2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H9ClN2O6S
CAS Number:
Molecular Weight:
356.74
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.25

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

[S](=O)(=O)(Nc2c(cccc2)C(=O)O)c1cc(c(cc1)Cl)[N+](=O)[O-]

InChI

1S/C13H9ClN2O6S/c14-10-6-5-8(7-12(10)16(19)20)23(21,22)15-11-4-2-1-3-9(11)13(17)18/h1-7,15H,(H,17,18)

InChI key

NJTHPOSQGFJTDP-UHFFFAOYSA-N

Biochem/physiol Actions

CTPI-2 is a selective mitochondrial citrate transporter (SLC25A1) inhibitor that displays H1299 anti-proliferation activity in a SLC25A1-dependent manner (IC50 <10  μM). When compared with CTPI-1, CTPI-2 exhibits 20-fold higher target affinity (hSLC25A1 KD = 3.5 μM vs. 63.6 μM, respectively) and is ~1000-fold more potent against H1299 sphere-forming capacity. CTPI-2 inhibits mitochondrial respiration and formation of patient NSCLC tumors-derived CSCs spheres in cultures (10-50 μM), as well as suppresses the growth of patent-derived NSCLC exnografts in mice in vivo (28 mg/kg q.o.d. i.p.).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Terumasa Umemoto et al.
The EMBO journal, 41(8), e109463-e109463 (2022-03-02)
In order to support bone marrow regeneration after myeloablation, hematopoietic stem cells (HSCs) actively divide to provide both stem and progenitor cells. However, the mechanisms regulating HSC function and cell fate choice during hematopoietic recovery remain unclear. We herein provide
Kexu Xiang et al.
Cell death & disease, 13(7), 641-641 (2022-07-23)
Oncogenic mutations in metabolic genes and associated oncometabolite accumulation support cancer progression but can also restrict cellular functions needed to cope with DNA damage. For example, gain-of-function mutations in isocitrate dehydrogenase (IDH) and the resulting accumulation of the oncometabolite D-2-hydroxyglutarate
Harvey R Fernandez et al.
Cell death and differentiation, 25(7), 1239-1258 (2018-04-14)
Therapy resistance represents a clinical challenge for advanced non-small cell lung cancer (NSCLC), which still remains an incurable disease. There is growing evidence that cancer-initiating or cancer stem cells (CSCs) provide a reservoir of slow-growing dormant populations of cells with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service