Skip to Content
Merck
All Photos(1)

Key Documents

SML3620

Sigma-Aldrich

UNC2250

≥98% (HPLC)

Synonym(s):

UNC 2250, UNC-2250, trans-4-((2-(Butylamino)-5-(5-(morpholinomethyl)pyridin-2-yl)pyrimidin-4-yl)amino)cyclohexanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H36N6O2
CAS Number:
Molecular Weight:
440.58
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.25

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

-10 to -25°C

SMILES string

N4(CCOCC4)Cc1cnc(cc1)c2c(nc(nc2)NCCCC)N[C@@H]3CC[C@H](CC3)O

InChI key

HSYSSKFCQHXOBP-MXVIHJGJSA-N

Biochem/physiol Actions

Potent and highly selective Mer receptor tyrosine kinase (MerTK) inhibitor in vitro and in vivo.
UNC2250 is a potent and highly selective Mer receptor tyrosine kinase inhibitor (MerTK IC50 = 1.7 nM; Tyro3/Axl IC50 = 100/270 nM) that reduces endogenous Mer phosphorylation level (697 B-ALL IC50 = 9.8 nM post 1h treatment) and blocks ligand-stimulated activation of a chimeric EGFR-Mer in cells. UNC2250 inhibits colony-forming potential in rhabdoid (by 36% at 100 nM; BT-12 cells) and NSCLC tumor cells (Colo699 IC50 = 100 nM) with good pharmacokinetic properties. UNC2250 is reported to reduce mortality in a murine LPS-induced acute lung injury (ALI) model (2 mg/kg i.v. )

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuanfeng Du et al.
Brain research, 1766, 147525-147525 (2021-05-20)
The NLR family pyrin domain-containing 3 (NLRP3) multiprotein complex is associated with neuroinflammation and poor prognosis after subarachnoid hemorrhage (SAH). Accumulating evidence shows that Mer tyrosine kinase (MerTK) alleviates inflammatory responses via a negative feedback mechanism. However, the contribution and
The suppressive effects of Mer inhibition on inflammatory responses in the pathogenesis of LPS-induced ALI/ARDS
Science Signaling, 15(724) (2022)
Weihe Zhang et al.
Journal of medicinal chemistry, 56(23), 9683-9692 (2013-11-08)
Abnormal activation or overexpression of Mer receptor tyrosine kinase has been implicated in survival signaling and chemoresistance in many human cancers. Consequently, Mer is a promising novel cancer therapeutic target. A structure-based drug design approach using a pseudo-ring replacement strategy

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service