Skip to Content
Merck
All Photos(1)

Key Documents

SML3513

Sigma-Aldrich

Lorcaserin hydrochloride

≥98% (HPLC)

Synonym(s):

(1R)-8-Chloro-2,3,4,5-tetrahydro-1-methyl-1H-3-benzazepine hydrochloride, (R)-8-Chloro-1-methyl-2,3,4,5-tetrahydro-1H-3-benzazepine hydrochloride, (R)-8-Chloro-1-methyl-2,3,4,5-tetrahydro-1H-benzo[d]azepine hydrochloride, APD 356 hydrochloride, APD-356 hydrochloride, APD356 hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H14ClN·HCl
CAS Number:
Molecular Weight:
232.15
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.04

Quality Level

Assay

≥98% (HPLC)

form

powder

drug control

USDEA Schedule IV

storage condition

desiccated

color

white to beige

solubility

H2O: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

Lorcaserin is an orally active, potent and selective 5-hydroxytryptamine receptor (5-HT2C) agonist (human/rat Ki = 15 /29 nM) with 18- and 104-fold higher potency than 5-HT2A and 5-HT2B by cellular inositol phosphate accumulation assay (EC50 = 9, 168, 943 nM, respectively) and good selectivity over 5-HT1A/3/4C/5A/6/7 and a panel of 67 other GPCRs and ion channels. Chronic daily treatment to rats (4.5-18 mg/kg p.o. b.i.d.) maintained on a high fat diet causes reductions in food intake and body weight gain.
Orally active, potent and selective 5-hydroxytryptamine receptor (5-HT2C) agonist in vitro and in vivo.

Caution

Hygroscopic

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lisa R Gerak et al.
Drug and alcohol dependence, 227, 108991-108991 (2021-09-06)
The opioid epidemic continues despite the availability of medications, including buprenorphine, for opioid use disorder (OUD); identifying novel and effective treatments is critical for decreasing the prevalence of OUD and ending this crisis. Buprenorphine alone does not markedly attenuate abuse-related
Yang He et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 41(26), 5734-5746 (2021-05-26)
Obesity is a serious global health problem because of its increasing prevalence and comorbidities, but its treatments are limited. The serotonin 2C receptor (5-HT2CR), a G-protein-coupled receptor, activates proopiomelanocortin (POMC) neurons in the arcuate nucleus of hypothalamus (ARH) to reduce
William J Thomsen et al.
The Journal of pharmacology and experimental therapeutics, 325(2), 577-587 (2008-02-07)
5-Hydroxytryptamine (5-HT)(2C) receptor agonists hold promise for the treatment of obesity. In this study, we describe the in vitro and in vivo characteristics of lorcaserin [(1R)-8-chloro-2,3,4,5-tetrahydro-1-methyl-1H-3 benzazepine], a selective, high affinity 5-HT(2C) full agonist. Lorcaserin bound to human and rat
Chronic Lorcaserin Treatment Reverses the Nicotine Withdrawal-Induced Disruptions to Behavior and Maturation in Developing Neurons in the Hippocampus of Rats
International Journal of Molecular Sciences, 22(2), 868-868 (2021)
Tanishka S Saraf et al.
Epilepsy research, 175, 106677-106677 (2021-06-16)
Recent preclinical and clinical studies suggest that lorcaserin, a preferential serotonin 2C receptor (5-HT2CR) agonist that was approved for the treatment of obesity, possesses antiepileptic properties. Here, we tested whether lorcaserin (1, 3, 5.6, 10 mg/kg) is prophylactic against audiogenic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service