Skip to Content
Merck
All Photos(1)

Key Documents

SML3478

Sigma-Aldrich

PNU-159682

≥95% (HPLC)

Synonym(s):

(8S,10S)-7,8,9,10-Tetrahydro-6,8,11-trihydroxy-8-(2-hydroxyacetyl)-1-methoxy-10-[[(1S,3R,4aS,9S,9aR,10aS)-octahydro-9-methoxy-1-methyl-1H-pyrano[4′,3′:4,5]oxazolo[2,3-c][1,4]oxazin-3-yl]oxy]-5,12-naphthacenedione, PNU 159682

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H35NO13
CAS Number:
Molecular Weight:
641.62
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

powder

storage condition

under inert gas

color

, Faint red to very dark red

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

PNU-159682, an active nemorubicin metabolite, is a highly potent anthracycline toxin that retains antitumor activity in vivo. Similarly to other anthracyclines PNU-159682 intercalates between the CG base pairs of DNA, but the stability of the complexes is much higher.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Carc. 2 - Muta. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Luigi Quintieri et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 11(4), 1608-1617 (2005-03-05)
Nemorubicin (3'-deamino-3'-[2''(S)-methoxy-4''-morpholinyl]doxorubicin; MMDX) is an investigational drug currently in phase II/III clinical testing in hepatocellular carcinoma. A bioactivation product of MMDX, 3'-deamino-3'',4'-anhydro-[2''(S)-methoxy-3''(R)-oxy-4''-morpholinyl]doxorubicin (PNU-159682), has been recently identified in an incubate of the drug with NADPH-supplemented rat liver microsomes. The aims
Stefania Mazzini et al.
Bioorganic & medicinal chemistry, 20(24), 6979-6988 (2012-11-17)
The antitumor anthracycline nemorubicin is converted by human liver microsomes to a major metabolite, PNU-159682 (PNU), which was found to be much more potent than its parent drug toward cultured tumor cells and in vivo tumor models. The mechanism of
Nikolas Stefan et al.
Molecular cancer therapeutics, 16(5), 879-892 (2017-03-05)
Antibody-drug conjugates (ADC) are highly potent and specific antitumor drugs, combining the specific targeting of mAbs with the potency of small-molecule toxic payloads. ADCs generated by conventional chemical conjugation yield heterogeneous mixtures with variable pharmacokinetics, stability, safety, and efficacy profiles.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service